Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Simplify sampling with easy access
Brand: Thermo Scientific™ 840-303200
This item is not returnable.
View return policy
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Sample holder for film/filter accommodates filters/lenses up to 50 mm long, 80mm tall and 10mm thick. Comes assembled in a removable sample compartment liner. | |
Measuring transmitting solid samples such as glass or plastic panels, optical filters and specialty films on glass or plastic substrates. | |
Filter/lenses up to 50 mm long x 80 mm tall x 10 mm thick. Minimum filter size 18 mm. |
Sample Holder for Film/Filter | |
GENESYS 140/150/180, BioMate 160 Spectrophotometers |