Learn More
Gilson™ Single Channel PIPETMAN™ M
GREENER_CHOICE
Single Channel PIPETMAN™ M is a Bluetooth™-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes.
Brand:  Gilson™ F81044
717.25 EUR valid until 2025-12-31
Use promo code "24312" to get your promotional price.
 View more versions of this product
Product Code. 16388147
  • 755.00 € / Each
Explore more special offers
Description
Description
  • Allows accurate and precise pipetting
  • The motorized piston allows for high pipetting reproducibility and accuracy in all modes
  • Associated with TRACKMAN™ Connected and its PipettePilot™ application, PIPETMAN™ M Connected reduces the risk of human error by transferring verifiable pipetting parameters and commands in the pipette
  • Increased functionality
  • The intuitive interface has five pipetting modes for a large number of applications: pipette, repetitive, mix, reverse, and custom
  • Create and transfer up to 10 custom protocols from PIPETMAN™ M Connected Software into your pipette
  • Bluetooth™ connection enables communication with TRACKMAN™ Connected and helps guide you through your protocol telling you where, when, and how much to pipette with real-time interactions
  • Simple and comfortable to use
  • Aspirate and dispense with one click of the push button and eject tips with ease
  • The lightweight and ergonomic design rests comfortably in your hand for maximum pipetting comfort
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Universal
Autoclavable
Reusable
Polypropylene
1
Yes
Pipette
Pipette Tips, TRACKMAN™ Connected
12.75 in.
8 in.
6.4 cm
Certified Quality
No
Yellow
Pipette Tip, TRACKMAN™ Connected
P300M BT
Pipetman M
Non-sterile
20 to 300 μL
1 Year
32.4 cm
2.5 in.
3.7 V and 420 mAh (1.55 Wh)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gilson™ Single Channel PIPETMAN™ M > Single Channel PIPETMAN™ M; Volume: 20 to 300 μL