Learn More
Manutan Single cutting station, cutting width 1250mm, roll weight 200kg
Brand: Manutan A016030

Description
An ergonomic packaging station especially suitable for lightweight film and corrugated board. Safe bi-directional cutting with adjustable take off height and roller bearing spindle. Each system is supplied with two cartridges, one circular blade, one flat blade. Ergonomically designed cutter cartridge easily replaced in a single operation.
- Single cutting station with one roll
- Cutting width: 1250mm
- Roll weight: 200kg
- Dimensions (H x W x D): 1060mm x 1590mm x 970mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.