missing translation for 'onlineSavingsMsg'
Learn More

Manutan Single wall box, 127mm x 127mm x 127mm

Brand:  Manutan A329266

Encompass_Preferred

Product Code. 12885428

  • 19.53 € / Pack of 25
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The cost-effective solution for packing less fragile items, economical and ideal for every day use. Strong, single wall boxes with brown kraft finish. Supplied flat for ease of storage.
  • Internal dimensions (L x H): 127mm x 127mm
  • Load capacity: 2kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers