missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Single wall box, 305mm x 305mm x 305mm
Brand: Manutan A329289
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
The cost-effective solution for packing less fragile items, economical and ideal for every day use. Strong, single wall boxes with brown kraft finish. Supplied flat for ease of storage.- Internal dimensions (L x H): 305mm x 305mm
- Load capacity: 10kg
Specifications
305 x 305 x 305 mm |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction