missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Single Well High Profile Reagent Reservoirs
Manufactured with virgin polypropylene for excellent chemical resistance
302.05 € - 356.00 €
Specifications
Certifications/Compliance | ANSI/SLAS, RNase/DNase-free, non-pyrogenic |
---|---|
Color | Clear |
Material | Polypropylene |
Type | Reagent Reservoir |
Disposable | Single-use |
Product Code | Brand | Capacity (Metric) | Sterility | Channels | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Sterility | Channels | Price | Quantity & Availability | |||
11320275
|
Axygen™
RES-SW384-HP-SI |
282 mL | Sterile | 384 |
354.00 €
Pack of 25 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
11310275
|
Axygen™
RES-SW384-HP |
282 mL | Non-sterile | 384 | N/A | ||||
11360275
|
Axygen™
RES-SW8-HP-SI |
290 mL | Sterile | 8 |
354.00 €
Pack of 25 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
11300275
|
Axygen™
RES-SW12-HP-SI |
290 mL | Sterile | 12 |
354.00 €
Pack of 25 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
11370275
|
Axygen™
RES-SW96-HP |
240 mL | Non-sterile | 96 |
340.00 €
Pack of 25 |
Estimated Shipment: 27-02-2025
Log in to see stock. |
|||
11350275
|
Axygen™
RES-SW8-HP |
290 mL | Non-sterile | 8 |
349.46 €
Pack of 25 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
11390265
|
Axygen™
RES-SW12-HP |
290 mL | Non-sterile | 12 |
349.46 €
Pack of 25 |
Estimated Shipment: 10-04-2025
Log in to see stock. |
|||
11380275
|
Axygen™
RES-SW96-HP-SI |
240 mL | Sterile | 96 |
356.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||
Description
Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.
- Durable and cost effective
- Various formats for multiple applications
Specifications
ANSI/SLAS, RNase/DNase-free, non-pyrogenic | |
Polypropylene | |
Single-use |
Clear | |
Reagent Reservoir |