missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Single Well Low Profile Reagent Reservoirs
GREENER_CHOICE

Product Code. p-4909858 Shop All Axygen Scientific Products
Click to view available options
Capacity (Metric):
86 mL
90 mL
92 mL
Sterility:
Non-sterile
Sterile
Channels:
1
96
384
Packaging:
25/Cs.
Individually Wrapped; 25/Cs.
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11301974

Brand: Axygen™ RESSW96LP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured with virgin polypropylene for excellent chemical resistance

Corning offers a complete line of Axygen single-well and multiple-well reagent reservoirs which are available in multiple formats for automated and manual use. All reservoirs are designed to comply with standard ANSI/SLAS footprint dimensions for microplates and are compatible with most automation workstations. Reagent reservoirs are manufactured with polypropylene for excellent chemical resistance.

  • Durable and cost effective
  • Various formats for multiple applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI/SLAS, RNase/DNase-free, non-pyrogenic
Color Clear
Capacity (Metric) 86 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
Packaging 25/Cs.
Channels 96
Disposable Single-use
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.