missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SiteClick™ Antibody Labeling Kits

Generate brightly labeled antibodies for flow cytometry, fluorescence imaging, or western blot detection using SiteClick™ Qdot™ R-PE, and Biotin Antibody Labeling Kits.

514.00 € - 614.00 €

Specifications

Product Type Labeling Kit
Content And Storage Store the entire kit at 2 to 6°C upon receipt
Labeling Method SiteClick Labeling System
Shipping Condition Wet Ice
View More Specs

Products 10
Product Code Brand Label or Dye Excitation/Emission Labeling Scale Price Quantity & Availability  
Product Code Brand Label or Dye Excitation/Emission Labeling Scale Price Quantity & Availability  
15138735
View Documents
Invitrogen™
S10453
Qdot 655 405/655 100 μg
600.00 €
Each
In Stock
to see stock.
Add to basket
 
15108745
View Documents
Invitrogen™
S10467
R-PE (R-Phycoerythrin) 496, 546, 565/578 100 μg
614.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15128735
View Documents
Invitrogen™
S10449
Qdot 525 405/525 100 μg
592.00 €
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
15136585
View Documents
Invitrogen™
S10454
Qdot 705 405/705 100 μg
600.00 €
1 set
In Stock
to see stock.
Add to basket
 
15148735
View Documents
Invitrogen™
S10450
Qdot 565 405/565 100 μg
598.00 €
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
15158735
View Documents
Invitrogen™
S10451
Qdot 585 405/585 100 μg
606.00 €
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
15168735
View Documents
Invitrogen™
S10452
Qdot 625 405/625 100 μg
606.00 €
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
15188735
View Documents
Invitrogen™
S10455
Qdot 800 100 μg
604.00 €
Each
In Stock
to see stock.
Add to basket
 
15198735
View Documents
Invitrogen™
S10469
Qdot 605 405/605 100 μg
514.00 €
Each
Estimated Shipment: 12-12-2024
to see stock.
Add to basket
 
15782966
View Documents
Invitrogen™
S20033
Biotin 100 to 250 μg
588.00 €
1 set
In Stock
to see stock.
Add to basket
 
Description

Description

Generate perfectly labeled antibodies with the SiteClick R-PE, Qdot, or Biotin Antibody Labeling Kits. Site selectivity is achieved by targeting the carbohydrate domains present on essentially all IgG antibodies, regardless of isotype and host species. Depending upon the label, the resulting SiteClick R-PE, Qdot, or Biotin conjugated antibody can be used in flow cytometry, fluorescence imaging, or Western blot detection. Each kit contains everything needed to label 100 μg of IgG antibody with an easy-to-follow, step-by-step protocol. Achieve highly efficient, site-specific, and reproducible labeling with the SiteClick R-PE, Qdot, or Biotin Antibody Labeling Kits.

Conventional amine or thiol labeling chemistry can result in labeling of the antigen binding domain or compromise the structure of an antibody. SiteClick labeling technology, however, attaches the label under mild conditions to the heavy chains of an IgG antibody, ensuring that the antigen binding domains remain available for binding to your antigen target. Because SiteClick Labeling Kits allow simple and gentle site-selective attachment of detection molecules to heavy chain N-linked glycans — far from the antigen-binding domain — excellent reproducibility is achieved during sequential labelings, as well from antibody to antibody. No harsh reduction steps are required, and antibody labeling is consistent and reproducible each time it is performed.

The SiteClick R-PE, Qdot, or Biotin Antibody labeling workflow consists of three steps:

1. antibody carbohydrate domain modification,
2. azide attachment to the antibody and
3. conjugation with the DIBO-modified label.

This workflow employs copper-free click chemistry to covalently link the label containing the DIBO moiety with the azide-modified antibody. Labeling antibodies with the SiteClick antibody labeling system gives you confidence that the antibody will be labeled the same way every time, with no need for reaction optimization. This means the same number of dyes are attached to each molecule, the antigen binding site is preserved, and the same results are achieved every time.

Important features of the SiteClick Qdot Antibody Labeling Kits:

• Include everything required to label 100 μg of IgG antibody (100–250 μg of IgG antibody with the SiteClick Biotin Antibody Labeling Kit)
• Easy to follow step-by-step protocol that require only a few hours hands-on time to complete.
• Highly efficient, site-specific, reproducible labeling chemistry results in high quality antibody conjugates.

Specifications

Specifications

Labeling Kit
Store the entire kit at 2 to 6°C upon receipt
SiteClick Labeling System
Wet Ice
Videos
SDS
Documents

Documents

Product Certifications