missing translation for 'onlineSavingsMsg'
Learn More

Sivelestat sodium salt, Tocris Bioscience™

Product Code. 18065992
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective leukocyte elastase inhibitor

Selective leukocyte elastase inhibitor (IC50 = 44 nM) that displays no activity at a range of other proteases. Inhibits NF-κB activation and LTB4-induced neutrophil transmigration in vitro. Significantly attenuates ischemia-induced spinal cord injury, decreases serum cytokine levels and reduces acute inflammatory lung injury in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150374-95-1
Molecular Formula C20H21N2NaO7S
Molecular Weight (g/mol) 456.445
InChI Key ZAIFANJZUGNYCK-UHFFFAOYSA-M
Synonym sivelestat sodium, sivelestat sodium salt, unii-0cll4232kd, sivelestat sodium salt hydrate, ono-5046.na, sivelestat sodium anhydrous, sivelestatsodium, c20h21n2o7s.na, glycine, n-2-4-2,2-dimethyl-1-oxopropoxy phenyl sulfonyl amino benzoyl-, monosodium salt
PubChem CID 23664980
IUPAC Name sodium;2-[[2-[[4-(2,2-dimethylpropanoyloxy)phenyl]sulfonylamino]benzoyl]amino]acetate
SMILES CC(C)(C)C(=O)OC1=CC=C(C=C1)S(=O)(=O)NC2=CC=CC=C2C(=O)NCC(=O)[O-].[Na+]

Specifications

Quantity 10 mg
Formula Weight 456.44
Percent Purity >98%
Chemical Name or Material Sivelestat sodium salt
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.