missing translation for 'onlineSavingsMsg'
Learn More
SKF 83566 hydrobromide, Tocris Bioscience™ Product Code.: 18004272

SKF 83566 hydrobromide, Tocris Bioscience™

Product Code. 18004272
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18004272

Brand: Tocris Bioscience 1586/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective D1-like antagonist

Potent and selective D1-like dopamine receptor antagonist (Ki∽ 0.56 nM for D1; KB = 2 μM for D2). Also antagonist at the vascular 5-HT2 receptor (Ki = 11 nM). Displays selective inhibition of adenylyl cyclase 2 (AC2); inactive against AC1 or AC5. Centrally active following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108179-91-5
Molecular Formula C17H19Br2NO
Molecular Weight (g/mol) 413.153
InChI Key SDQJYYGODYRPBR-UHFFFAOYSA-N
Synonym skf 83566 hydrobromide, 8-bromo-3-methyl-5-phenyl-2,3,4,5-tetrahydro-1h-benzo d azepin-7-ol hydrobromide, 8-bromo-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1h-3-benzazepin-7-ol hydrobromide, 8-bromo-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1h-3-benzazepin-7-olhydrobromide, 8-bromo-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol hydrobromide, 8-bromo-3-methyl-5-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepin-7-ol hydrobromide
PubChem CID 23581817
IUPAC Name 8-bromo-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol;hydrobromide
SMILES CN1CCC2=CC(=C(C=C2C(C1)C3=CC=CC=C3)O)Br.Br

Specifications

Quantity 10 mg
Formula Weight 413.15
Percent Purity >98%
Chemical Name or Material SKF 83566 hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SKF 83566 hydrobromide, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.