missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
High affinity, selective dopamine D1-like receptor agonist. Ki values are 3.2, 3.1, 186, 66, 335, 1167, 1251 and 1385 nM at recombinant D1, D5, D2, D3, D4, 5-HT2A, α1A and α1B receptors respectively. Stimulates adenylyl cyclase (EC50 = 65 nM) but not phosphoinositide hydrolysis. Induces extreme arousal and hyperlocomotion following subcutaneous administration in monkeys.

Specifications
Specifications
Quantity | 10 mg |
Synonym | skf 83822 hydrobromide, skf-83822 hydrobromide, 6-chloro-2,3,4,5-tetrahydro-1-3-methylphenyl-3-2-propenyl-1h-3-benzazepine-7,8-diol hydrobromide, n-allyl-6-chloro-1-3-methylphenyl-2,3,4,5-tetrahydro-3-benzazepine-7,8-diol hydrobromide, 3-allyl-6-chloro-1-3-methylphenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, 3-allyl-6-chloro-7,8-dihydroxy-1-3-methylphenyl-2,3,4,5-tetrahydro-1h-3-benzazepinium bromide, 6-chloro-1-3-methylphenyl-3-prop-2-en-1-yl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, 6-chloro-7,8-dihydroxy-1-3-methylphenyl-3-prop-2-en-1-yl-2,3,4,5-tetrahydro-1h-3-benzazepinium bromide, skf83822 hydrobromide, 1h-3-benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-1-3-methylphenyl-3-2-propen-1-yl-, hydrobromide 1:1 |
Formula Weight | 424.76 |
Percent Purity | >98% |
Chemical Name or Material | SKF 83822 hydrobromide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SKF 83822 hydrobromide, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction