missing translation for 'onlineSavingsMsg'
Learn More
SKF 83959 hydrobromide, Tocris Bioscience™ Product Code.: 18075092

SKF 83959 hydrobromide, Tocris Bioscience™

Product Code. 18075092
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18075092

Brand: Tocris Bioscience 2074/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

D1-like partial agonist

Dopamine D1-like receptor partial agonist (Ki values are 1.18, 7.56, 920 and 399 nM for rat D1, D5, D2 and D3 receptors respectively). May act as an antagonist in vivo, producing anti-Parkinsonian effects and antagonizing the behavioral effects of cocaine. Shown to potentiate agonist binding of the σ1 receptor.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67287-95-0
Molecular Formula C18H21BrClNO2
Molecular Weight (g/mol) 398.725
InChI Key FHYWNBUFNGHNCP-UHFFFAOYSA-N
Synonym skf 83959 hydrobromide, skf-83959 hydrobromide, 6-chloro-7,8-dihydroxy-3-methyl-1-3-methylphenyl-2,3,4,5-tetrahydro-1h-3-benzazepine hydrobromide, 6-chloro-3-methyl-1-3-methylphenyl-2,3,4,5-tetrahydro-1h-3-benzazepine-7,8-diol hydrobromide, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1-3-methylphenyl-1h-3-benzazepine-7,8-diol hydrobromide, 1h-3-benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1-3-methylphenyl-, hydrobromide, 1h-3-benzazepine-7,8-diol, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1-3-methylphenyl-, hydrobromide 1:1, skf-83959 hydrobromide hplc , solid, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1-3-methylphenyl-1h-3-benzazepine-7,8-diol, 6-chloro-7,8-dihydroxy-3-methyl-1-3-methylphenyl-2,3,4,5-tetrahydro-1h-3-benzazepinium bromide
PubChem CID 11957685
ChEBI CHEBI:63996
IUPAC Name 9-chloro-3-methyl-5-(3-methylphenyl)-1,2,4,5-tetrahydro-3-benzazepine-7,8-diol;hydrobromide
SMILES CC1=CC=CC(=C1)C2CN(CCC3=C(C(=C(C=C23)O)O)Cl)C.Br

Specifications

Quantity 10 mg
Formula Weight 398.73
Percent Purity >98%
Chemical Name or Material SKF 83959 hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SKF 83959 hydrobromide, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.