Learn More
Notrax Skywalker HD™ ESD Nitrile FR Rubber Tiles with Bubble Pattern
Heavy duty rubber tiles can be easily assembled for coverage of large areas or individual workstations. Ergonomic benefit derived from a 13 mm thick worker platform with a bubble pattern for maximum fatigue relief in dry industrial environments.
Brand:  Notrax 464S0033BL
Product Code. 17782148
  • 292.00 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Heavy duty rubber tiles of 91 x 91 cm can be easily assembled for coverage of large areas or individual work stations
  • Ergonomic benefit derived from a 13 mm thick worker platform with a bubble pattern for maximum fatigue relief in dry industrial environments
  • A tooled grid of 30 x 30 cm sections on the underside of the mat makes further customization quick and easy
  • Attachable bevelled edges prevent tripping and allow inside and outside corners
  • Slip resistance rating R9 according to DIN 51130
  • Static dissipative natural rubber compound Meets IEC61340-4-1 (category DIF), measured resistance Rg 106 to 109 Ω, Rg 106 to 109 Ω
  • Fire classification Bfl-S1 tested according to EN 13501-1
  • Resistant to most industrial oils and suitable for welding areas
Specifications
Specifications
Black
91 cm
Industrial and Commercial Matting
36 in.
ESD and Fire Retardant Nitrile Rubber Compound
91 cm