missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Under Bench Cabinet for Acids and Alkalis SL-CLASSIC-UB
Under bench cabinet for acids and alkalis with pull out shelf.
745.00 € - 1009.95 €
Specifications
Depth (Metric) Interior | 531 mm |
---|---|
Color | Light Gray |
Product Type | Under bench cabinet for acids and alkalis |
For Use With (Application) | Safe and approved storage of aggressive, non flammable hazardous materials in working areas |
Height (Metric) Exterior | 600 mm |
Alert:
For indoor use only
Product Code | Brand | Configuration | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Material | Price | Quantity & Availability | ||||
10766451
|
asecos™
30619-001-30621 |
4 x pull-out shelf (panel/PP) | Steel | N/A | |||||
15985696
|
asecos™
3062389230624 |
1 x pull-out shelf with PP-sump, load capacity: 25 kg, capacity: 11 liters (panel melamine resin-coated/PP) | Chipboard |
745.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12653515
|
asecos™
3062289230625 |
2 x pull-out shelf (panel melamine resin-coated/PP) | Chipboard |
889.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995696
|
asecos™
3062389230625 |
2 x pull-out shelf with PP-sump, load capacity: 25 kg, capacity: 11 liters (panel melamine resin-coated/PP) | Chipboard |
863.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Safe and approved storage of aggressive, non flammable hazardous materials in working areas
Function / construction
- Robust construction and longevity: made out of highly resistant melamin resin special plates
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- No unauthorised use: door lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Certificates
- type tested in accordance with
- EN 16121/16122 conformity
- GS certified
Specifications
531 mm | |
Under bench cabinet for acids and alkalis | |
600 mm |
Light Gray | |
Safe and approved storage of aggressive, non flammable hazardous materials in working areas |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title