Learn More
SLC22A9 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27998-1-AP-150UL
 View more versions of this product
Product Code. 16875945
  • 410.00 € / 150µL
Explore more special offers
Description
Description
The SLC22A9 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SLC22A9. This antibody recognizes human, mouse antigen. The SLC22A9 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SLC22A9
Polyclonal
Unconjugated
150 μL
114571
IgG
Western Blot, Immunohistochemistry
500 μg/mL
Rabbit
Primary
Human, Mouse