Learn More
Lenz Laborglasintrumente Sleeves with Grip Collar
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01101724
This item is not returnable.
View return policy
Description
- Outstanding chemical resistance at operating temperatures from -200 to +260 °C
- Sturdy design for permanent use
- With grip collar and sealing rings for excellent tightness
Specifications
PTFE | |
NS 24/29 |