Learn More
Ibidi μ-Slide 8 Well Collagen I
μ-Slide 8 Well Collagen I is a chambered coverslip with 8 wells.
Brand:  Ibidi 80829
Encompass_Preferred
Product Code. 17844834
  • 451.33 € / Pack of 15
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A chambered coverslip with 8 wells for cell culture, immunofluorescence, and high-end microscopy
  • bottom material: #1.5 Polymer Coverslip
  • Collagen I coated
  • sterilized
Specifications
Specifications
Microscope Slide
8
15