Learn More
Ibidi μ-Slide 8 Well high μ-Pattern RGD, cir100, pit500, hex Trial Pack
μ-Slide 8 Well high μ-Pattern RGD, cir100, pit500, hex Trial Pack is an 8 well chambered coverslip with micropatterned surface.
Brand: Ibidi 83802-S
This item is not returnable.
View return policy
Description
- A chambered coverslip with 8 wells and micropatterned surface for multi-cell assays with fluorescence microscopy readout
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
- sterilized
Specifications
Microscope Slide | |
8 | |
2 |