Learn More
Ibidi μ-Slide 8 Well high μ-Pattern RGD Test Patterns 3
μ-Slide 8 Well high μ-Pattern RGD Test Patterns 3 is an 8 well chambered coverslip with micropatterned surface.
Brand: Ibidi 83853
This item is not returnable.
View return policy
Description
- An 8 well chambered coverslip with micropatterned surface to test different patterns for the establishment of cell culture assays
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
- sterilized
Specifications
Microscope Slide | |
8 | |
2 |