missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Devices, 10K MWCO

Perform sample buffer exchange with these 10,000-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.

Brand:  Thermo Scientific™ 69570

 View more versions of this product

Product Code. 10185003

  • 266.00 € / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Dialysis Unit
10 kDa
MINI Dialysis Device
0.1 mL
50 Devices
Slide-A-Lyzer™
Cellulose Membrane, Dialysis
Buffer Exchange, Protein
Dialysis Units
Store at room temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK395331 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number69570
Lot NumberZJ387464 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number69570
Lot NumberZI387463 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number69570
Lot NumberZH386528 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number69570
Lot NumberZG386527 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number69570
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Devices, 10K MWCO > MINI Dialysis Device; Cup, Microcentrifuge; 50 Devices

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.