missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ MINI Dialysis Device, 7K MWCO

Product Code. p-4524602 Shop All Thermo Scientific Products
Click to view available options
Quantity:
250 Devices
50 Devices
Unit Size:
Pack of 250
Pack of 50
This item is not returnable. View return policy

Product Code. 10083213

Brand: Thermo Scientific™ 69560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform sample buffer exchange with these 7000-MW cut-off (MWCO) disposable polypropylene cups with integrated, low-binding membranes.

Slide-A-Lyzer MINI Dialysis Devices, 7K MWCO, are disposable polypropylene cups with integrated, low-binding membranes for dialysis and high recovery of proteins and macromolecules larger than 7,000 Daltons in volumes from 10 to 100 μL.

Slide-A-Lyzer MINI Dialysis Devices were developed as a convenient means for dialyzing small-volume samples of nucleic acids and proteins without undue sample loss. Low molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, micro conjugation reactions, and concentration can be accomplished with these devices. Slide-A-Lyzer MINI Dialysis Devices are manufactured in a HEPA filter/clean environment and auto-bagged after assembly to ensure that they are free of contaminants and ready to use.

Slide-A-Lyzer MINI Dialysis Devices allow easy sample addition and removal using a standard laboratory pipette and can be used for single or arrays of samples. Unlike other small sample separation devices, there is no equipment to assemble, no need for syringe adapters, and no laborious steps necessary to manipulate the small volume sample. The regenerated cellulose membrane is compatible with common chemicals and buffers.

The 0.1 mL MINI dialysis devices are designed to be capped (caps included) and inserted into standard 1.5 mL micro-centrifuge tubes. Alternatively, as many as 25 MINI devices can be placed in a Slide-A-Lyzer MINI Dialysis Device Float for dialysis of multiple samples inside a large beaker.

Features of Slide-A-Lyzer MINI Dialysis Devices, 7K MWCO:
Flexible format—enables user to dialyze a single sample (tube) or multiple samples (25 array)
Excellent sample recovery—low-binding plastic and small membrane surface area minimize sample loss compared to filtration and resin systems
One-step protocol—does not require laborious procedures or expensive equipment
100% leak-tested—innovative design does not permit 'wicking' that can occur in home-made devices
Time saving—performs desalting of nucleic acids and proteins (>95% recovery) in less than 15 minutes, which improves sample resolution, quantification, and conjugation

Using the 0.1 mL MINI Device, 100 μL of pH 2.8 glycine buffer can be equilibrated with 1 liter of pH 9.4 bicarbonate buffer in less than 10 minutes. As with all dialysis applications, the observed rate of dialysis for each sample used with these devices depends on a variety of factors including membrane to sample volume; temperature and agitation rates; the molecular weight, shape, degree of hydration, and charge of the molecules passing through the membranes; and the pH, ionic strength, and solvent characteristic of the buffers being used.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cup, Microcentrifuge
Product Line Slide-A-Lyzer
MWCO 7.0 kDa
Membrane Cellulose Membrane, Dialysis
Format MINI Dialysis Device
Description MINI Dialysis Device, 0.1 mL
Purification Target Buffer Exchange, Protein
Sample Volume (Metric) 0.1 mL
Quantity 50 Devices
Content And Storage Store at room temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.