missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Slide-A-Lyzer™ Dialysis Cassettes, 20K MWCO

Product Code. p-7226481 Shop All Thermo Scientific Products
Click to view available options
Sample Volume (Metric):
0.5 mL
3 mL
12 mL
30 mL
Unit Size:
Pack of 10
Pack of 6
Pack of 8
This item is not returnable. View return policy

Product Code. 10668884

Brand: Thermo Scientific™ 66005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use this 20K MWCO semi-permeable membrane to remove salts and contaminants with maximal sample recovery.

Slide-A-Lyzer Dialysis Cassettes (20K MWCO) facilitate the simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules larger than 20,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange, and more complete sample recovery.

Slide-A-Lyzer Dialysis Cassettes (20K MWCO) facilitate the simple and effective removal of buffer salts and small contaminants from proteins and other macromolecules larger than 20,000 Daltons. They are available in four sizes to accommodate sample volumes between 0.5 and 30 mL. These devices are easier to use than traditional dialysis tubing, providing faster buffer exchange and more complete sample recovery. Slide-A-Lyzer cassettes can be used for a wide range of applications including low-molecular weight contaminant removal, buffer exchange, desalting, equilibrium dialysis, and sample concentration. Liquid samples are easily added and removed by penetrating the self-sealing gasket with a hypodermic needle attached to a syringe. No knots, clips, or caps are needed to seal the units and prevent leaking and sample-loss during the dialysis procedure.

Slide-A-Lyzer Dialysis Cassettes are constructed from two sheets of low-binding, regenerated-cellulose dialysis membrane that are hermetically sealed on either side of a silicone-like gasket inside an inert plastic frame. The membrane and cassette materials are compatible with most common laboratory chemicals and buffers.

Features of Slide-A-Lyzer Dialysis Cassettes:
Easy to use—no knots or clamps are needed; just inject sample into cassette and begin dialysis
Fast dialysis—flat cassette chamber with two membranes provides high surface-area to volume ratio that maximizes diffusion rate compared to cylindrical dialysis tubing
High recovery—rectangular cassette design maximizes recovery of entire sample volume via any one of the four corner injection ports
Four cassette sizes—select the cassette that best suits your sample volume
Color-coded—purple cassette frame specifies cassettes with the 20,000 MW pore size membrane

Membrane specifications:
Composition: regenerated cellulose synthesized by the Viscose method
Thickness: 0.9 to 1.2 mil (22.5 to 30 μm)
Hydration time required before use: 2 minutes
Glycerol content: none
Sulfur content: 0.04%
Heavy metals content: trace

Volume ranges:
• 0.5 mL = 0.1 to 0.5 mL
• 3 mL = 0.5 to 3 mL
• 12 mL = 3 to 12 mL
• 30 mL = 12 to 30 mL

TRUSTED_SUSTAINABILITY

Specifications

Description Slide-A-Lyzer Dialysis Cassettes, 20K MWCO, 0.5mL
Content And Storage Store at room temperature
Purification Target Buffer Exchange, Protein
Membrane Cellulose Membrane, Dialysis
Format Cassette
MWCO 20 kDa
Product Line Slide-A-Lyzer
Quantity 10 Cassettes
Sample Volume (Metric) 0.5 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.