Learn More
Ibidi μ-Slide Chemotaxis ibiTreat
μ-Slide Chemotaxis ibiTreat is a ready-to-use microscopy slide for chemotaxis assays.
Brand: Ibidi 80326
This item is not returnable.
View return policy
Description
- A ready-to-use microscopy slide for chemotaxis assays with fast or slow migrating cells
- suitable for 2D or 3D cell culture
- stable chemotactic gradients for long-term experiments
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Microscope Slide | |
10 |