Learn More
Ibidi μ-Slide I 0.4 Luer Uncoated
A μ-Slide has one channel for applying defined shear stress.
Brand: Ibidi 80171
This item is not returnable.
View return policy
Description
- A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
- easy to connect with tubing and pump systems via female Luers
- channel height: 400 μm
- bottom material: #1.5 Polymer Coverslip
- hydrophobic
- sterilized
Specifications
Microscope Slide | |
15 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ibidi μ-Slide I 0.4 Luer Uncoated
Spot an opportunity for improvement?Share a Content Correction