Learn More
Ibidi μ-Slide I 0.6 Luer ibiTreat
A μ-Slide has one channel for applying defined shear stress.
Brand: Ibidi 80186
This item is not returnable.
View return policy
Description
- A μ-Slide with one channel for cell cultivation under flow and for applying defined shear stress
- easy to connect with tubing and pump systems via female Luers
- channel height: 600 μm
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Microscope Slide | |
15 |