Learn More
Ibidi μ-Slide I ibiTreat
A μ-Slide has one channel for imaging inside a channel.
Brand:  Ibidi 80106
Encompass
Product Code. 17256143
  • 282.00 € / Pack of 15
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A combination of a cell culture chamber and a coverslip for imaging inside a channel
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Specifications
Microscope Slide
15