Learn More
Ibidi μ-Slide I Luer 3D Uncoated
A μ-Slide has one channel and three wells.
Brand:  Ibidi 87171
Encompass
Product Code. 17236263
  • 417.00 € / Pack of 15
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A μ-Slide with one channel and three wells for culturing cells on a 3D gel matrix with defined flow
  • easy to connect with tubing and pump systems via female Luers
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Specifications
Microscope Slide
15