Learn More
Ibidi μ-Slide I Luer Variety Pack ibiTreat
μ-Slide I Luer Variety Pack ibiTreat includes a variety of μ-Slide I Luer channel slides.
Brand: Ibidi 81106
This item is not returnable.
View return policy
Description
- Variety of μ-Slide I Luer channel slides with 4 times each channel height
- specially suited for optimizing experimental conditions in flow applications
- easy to connect with tubing and pump systems via female Luers
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Microscope Slide | |
16 |