Learn More
Ibidi μ-Slide Spheroid Perfusion Uncoated
μ-Slide Spheroid Perfusion Uncoated is a perfusable channel slide with 3 x 7 wells.
Brand: Ibidi 80351
This item is not returnable.
View return policy
Description
- A perfusable channel slide with 3 x 7 wells for high-end microscopy
- easy to connect with tubing and pump systems via female Luers
- bottom material: #1.5 Polymer Coverslip
- hydrophobic
- sterilized
Specifications
Microscope Slide | |
15 |