Learn More
Kartell Slide Storing System
Slide storing system designed for the classification and storage of microscope slides, made of high impact PMMA. The semi-transparent ice color gives an attractive appearance while the metal structure makes it stable, while ensuring the value of lightness, functionality, strength and durability.
Brand:  Kartell 00945L1
 View more versions of this product
Product Code. 18087761
  • 811.00 € / Each
Explore more special offers
Description
Description
  • Each single drawer can hold up to 13 supports for slides (code 921) and transparent cover plates (code 920), to be ordered separately
  • Each slides support can contain 100 microscope slides, easily extractable
  • Each cavity is numbered
  • The transparent lids guarantee an easy check of the contents, protecting the slides from dust and other foreign bodies
  • The complete Storing System can therefore contain up to 5,200 slide
Specifications
Specifications
Slide Storage Cabinet
5000
Classification and storage of microscope slides
4 Swivel Castors