Learn More
Ibidi μ-Slide VI 0.1 ibiTreat
μ-Slide VI 0.1 ibiTreat is a 6 channel slide for parallel flow assays and immunofluorescence.
Brand: Ibidi 80666
This item is not returnable.
View return policy
Description
- A 6 channel slide for parallel flow assays and immunofluorescence that uses a minimal amount of reagents and medium
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Microscope Slide | |
15 |