Learn More
Ibidi μ-Slide VI 0.1 Uncoated
μ-Slide VI 0.1 Uncoated is a 6 channel slide for parallel flow assays and immunofluorescence.
Brand:  Ibidi 80661
Encompass
Product Code. 17276193
  • 349.50 € / Pack of 15
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A 6 channel slide for parallel flow assays and immunofluorescence that uses a minimal amount of reagents and medium
  • bottom material: #1.5 Polymer Coverslip
  • hydrophobic
  • sterilized
Specifications
Specifications
Microscope Slide
15