Learn More
Ibidi μ-Slide VI 0.4 Bioinert
μ-Slide VI 0.4 Bioinert is a 6 channel slide with a non-adherent surface.
Brand: Ibidi 80600
This item is not returnable.
View return policy
Description
- A 6 channel slide with a non-adherent surface for flow experiments and fluorescence microscopy
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert)
- sterilized
Specifications
Microscope Slide | |
15 |