Learn More
Ibidi μ-Slide VI 0.4 Collagen IV
μ-Slide VI 0.4 Collagen IV is a 6 channel slide for parallel flow assays and immunofluorescence.
Brand: Ibidi 80602
This item is not returnable.
View return policy
Description
- A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
- bottom material: #1.5 Polymer Coverslip
- Collagen IV coated
- sterilized
Specifications
Microscope Slide | |
15 |