Learn More
Ibidi μ-Slide VI 0.4 ibiTreat, Bulk Pack
μ-Slide VI 0.4 ibiTreat, Bulk Pack is a 6 channel slide for parallel flow assays and immunofluorescence.
Brand:  Ibidi 80606-90
Encompass
Product Code. 17236193
  • 1627.50 € / Pack of 90
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Specifications
Microscope Slide
90