Learn More
Ibidi μ-Slide VI 0.4 ibiTreat
μ-Slide VI 0.4 ibiTreat is a 6 channel slide for parallel flow assays and immunofluorescence.
Brand:  Ibidi 80606
Encompass
Product Code. 16438933
  • 319.50 € / Pack of 15
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A 6 channel μ-Slide suitable for flow experiments and for immunofluorescence assays
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Specifications
Specifications
Microscope Slide
15