Learn More
Ibidi μ-Slide VI 0.4 μ-Pattern RGD, sqr30, pit110, hex
μ-Slide VI 0.4 μ-Pattern RGD, sqr30, pit110, hex is a 6 channel slide with micropatterned surface .
Brand:  Ibidi 83603
Encompass_Preferred
Product Code. 17874784
  • 774.40 € / Pack of 10
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • A 6 channel slide with micropatterned surface
  • bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert) and RGD pattern
  • sterilized
Specifications
Specifications
Microscope Slide
10