Learn More
Ibidi μ-Slide y-shaped ibiTreat
μ-Slide y-shaped ibiTreat is a channel slide to study blood vessel bifurcation.
Brand: Ibidi 80126
This item is not returnable.
View return policy
Description
- A channel slide with bifurcations of 30 and 45 degree for bifurcation studies and blood vessel simulation
- easy to connect with tubing and pump systems via female Luers
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Specifications
Microscope Slide | |
15 |