Learn More
Micronova™ SlideMop™ Flat Head Frame and Mop Covers
BIOPHARMAGREENER_CHOICE
A simple, effective mop system featuring sturdy, collapsible plates that easily slide into the quilted polyester mop cover and lock into place with an effortless downward push. Available in a choice of stainless steel or polypropylene frame.
Brand:  Micronova™ MZCR63-16
 View more versions of this product
Product Code. 16656172
  • 208.00 € / Pack of 24
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Micronova SlideMop™ Flat Head Mop Frame and Covers is designed for use in the traditional “dip and wring” method or as a “saturation” method where liquid disposal and/or space saving is a factor.
  • Collapsible flat head mop frame and polyester covers
  • Lightweight, economical design for use throughout the facility
  • Quilted polyester mop covers with polyester pad interiors for disinfectant dispersal
  • Choice of Stainless Steel or Plastic Frame
  • Swivel joint ensures correct lift and pull or figure 8 cleaning techniques
  • For use with standard plastic or aluminum threaded handles
  • Can be used with SSU Series Stainless Steel Handle with an ACME adapter
  • Use with W-94 or W-92 wringer
  • Manufactured for use in Controlled Environments, ISO 5 thru 9
  • Available gamma irradiated: 25 kGy minimum dose
  • Good chemical resistance
  • Autoclavable
Specifications
Specifications
SlideMop, NovaPoly, Smooth Polyester, Flat Head Slip-on Mop Cover
16 in.
41 cm
Reusable
Buckets and Wringer
Polyester with Polyester Fill
White
Non-sterile
Autoclavable
Item may be Laundered and Reused. Care should be taken to fully eliminate disinfectants prior to reuse or steam sterilization. Water or Dryer temperature should not exceed 180°C.
Cleaning and Sanitization of Cleanroom Surfaces