missing translation for 'onlineSavingsMsg'
Learn More

Horst™ Slim Heating Tape with High Specific Heat Output

Double-laid heating conductors in a slim, insulated tape.

Brand:  Horst™ 25204

 View more versions of this product

Product Code. 10468641

  • 180.00 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Slim design.
  • Electrically insulated with PTFE.
  • Metallic protective braid of nickel-plated copper.
Specifications

Specifications

Bending radius: 10 mm, Thickness: 4 mm
230 V
1 m
4 mm
3.0 m
PTFE, Nickel-plated Copper
150 W
250°C (Nominal)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers