Learn More
Micronova™ SlimLine™ Cleanroom Cart System
A durable, double or triple bucket cart system with an electropolished, stainless steel frame, optional wringer bar, and three grip-it holders for easy storage and interchange of handles and mop heads. Systems comes standard with color coded, 22.7 liter (6 gal.) polypropylene buckets.
3165.00 € - 3605.00 €
Specifications
Autoclavable Autoclavable
For Use With (Equipment) SlimLine™ Cleanroom Cart System
Material Stainless Steel
View More Specs
Products 2
Product Code Brand Product Type Color Price Quantity & Availability  
Product Code Brand Product Type Color Price Quantity & Availability  
11962455
View Documents
Micronova™
C-22S
Double Bucket Cart Blue, Yellow
3165.00 €
Each
Estimated Shipment: 18-03-2025
to see stock.
Add to basket
11972455
View Documents
Micronova™
C-33S
Triple Bucket Cart Blue, Yellow, Red
3605.00 €
Each
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
Description
Description
The Micronova SlimLine™ Stainless Steel Cart Systems include an electropolished stainless steel cart and come standard with either two or three, color-coded, polypropylene buckets to distinguish between clean and rinse buckets. The three grip-it holders at the back of the cart allow for easy interchange and storage of handles and mop heads. An optional wringer bar accommodates either the Downpress or Flat Head Wringers, allowing placement above the rinse bucket for simplified wringing.
  • Cart is crafted from electropolished stainless steel for durability and long life
  • Includes three grip-it holders at the back of the cart
  • Easy storage and interchange of handles and mop heads
  • Comes standard with color-coded, 6-gallon (22-L), polypropylene buckets
  • Double bucket system comes with 1 blue and 1 yellow bucket
  • Triple bucket system includes an additional red bucket
  • Optional wringer bar allows wringers to be placed above the rinse bucket
  • Buckets are best autoclaved in the inverted position
  • Entire unit is autoclavable
Specifications
Specifications
Autoclavable
SlimLine™ Cleanroom Cart System
Stainless Steel