Learn More
Manutan Sling hook for 10mm chain
Brand:  Manutan A341456
Encompass_Preferred
Product Code. 12858568
  • 99.46 € / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
A range of high strength chain and fittings to suit a variety of lifting, lashing, suspending or security applications. Fittings - 'connector' for linking components and/or chain consists of 2 matched halves joined by a cross pin and tension bush. Other fittings are connected to chain using the Kuplex clevis system with load retaining pin. Test certificate supplied
  • Sling hook
  • Chain diameter: 10mm
  • Workload Limit: 3200