missing translation for 'onlineSavingsMsg'
Learn More

3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600

Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.

Brand:  3M™ Safety-Walk™ 7100001946

 View more versions of this product

Product Code. 17108680

  • 252.00 € / 1 roll
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Anti-slip tape for light to heavy shoe-traffic areas
  • High quality abrasive particles to ensure long-lasting friction in any application
  • Highly durable surface withstands the rigors of equipment traffic
  • High performance, pressure-sensitive adhesive for easy application
  • Custom shapes and sizes are also available
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

18.3 m x 102 mm
Light to Heavy Traffic Areas
1 Ea.
102 mm
Transparent
Mineral-coated
18.3 m
4 in.
Pressure Sensitive Adhesive
Floor Tape
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.