missing translation for 'onlineSavingsMsg'
Learn More

3M™ Safety-Walk™ Slip Resistant General Purpose Tape, Series 600

Mineral coated, slip resistant material features a highly durable surface. Low profile design helps reduce tripping hazards.

Brand:  3M™ Safety-Walk™ 7100067526

 View more versions of this product

Product Code. 17158700

  • 252.00 € / Pack of 4
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Anti-slip tape for light to heavy shoe-traffic areas
  • High quality abrasive particles to ensure long-lasting friction in any application
  • Highly durable surface withstands the rigors of equipment traffic
  • High performance, pressure-sensitive adhesive for easy application
  • Custom shapes and sizes are also available
  • NFSI High Traction Certification
TRUSTED_SUSTAINABILITY
Specifications

Specifications

18.3 m x 25 mm
Light to Heavy Traffic Areas
18.3 m
1 in.
Pressure Sensitive Adhesive
Floor Tape
Mineral-coated
60 ft.
4 Pk.
25 mm
Black
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers