Learn More
Thermo Scientific™ SLIPFREE Ferrules
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
Brand:  Thermo Scientific™ 36024
 View more versions of this product
Product Code. 10140643
  • 23.16 € / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ SLIPFREE Ferrules are for use with SLIPFREE connectors for HPLC columns.
  • Vespel ferrules replace the standard Vespel ferrules supplied with SLIPFREE column connectors
  • Kel-F and PEEK are offered for applications in which Vespel is not suitable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sealing Ferrule
Vespel sealing ferrule
SLIPFREE Connectors for HPLC Columns
Vespel