missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SlowFade™ Glass Soft-Set Antifade Mountant
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
2 mL
5 x 2 mL
missing translation for 'unitSize'
2mL
Pack of 5
Beschreibung
- A glycerol-based, ready-to-use, non-curing mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides
- Formulated to have a refractive index of 1.52 for sharper images, and antifade components to resist photobleaching
- Results in sharp, bright images with almost any fluorophore and wide-field, confocal, or super-resolution microscopes, at focal depths of 0–500 μm
- Mountant is non-curing, allowing cells to be imaged immediately after mounting, without any of the adverse effects on cell morphology that a curing mountant might cause
- It comes ready to use - just apply a drop to the sample, add a coverslip, and image
- Available with or without DAPI nuclear stain
- Prolong Glass Antifade Mountant is recommended if a curing, hard-set mountant is needed, also with a refractive index of 1.52 for years of slide storage
Features of SlowFade Glass Antifade Mountant include:
- Ready-to-use, glycerol-based, non-curing antifade mountant
- Refractive index of 1.52, with minimized spherical aberrations
- Up to three times better axial resolution compared to regular mountant
- Visible tissue optical clearing, even in sections up to 1 mm thick
- Sharp, bright images at any focal depth from 0 to 500 μm
Spezifikation
Spezifikation
| Quantity | 2 mL |
| Product Type | Antifade Mountant |
| Content And Storage | Contains 1 dropper bottle containing 2 mL reagent |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
| Product Line | SlowFade |
| Reagent Type | Antifade Solution |
| Volume (Metric) | 2 mL |
For Research Use Only. Not for use in diagnostic procedures.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur