missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ SLX-CLASSIC Recirculating Air Safety Storage Cabinets
For the safe and approved storage of aggressive, non-flammable hazardous materials in workrooms.
Brand: Asecos™ 3293900130615
This item is not returnable.
View return policy
Description
- Robust construction and longevity – cabinet body made of powder-coated sheet steel with safety elements assembled outside the storage compartment for increased protection against corrosion
- Two-in-one – two hermetically separated storage compartments allow the isolated storage of acids and bases in one cabinet
- No unauthorized use – doors lockable with profile cylinder with integration in an existing locking system possible
- Easy alignment – adjusting aids to compensate for uneven floors
- Recirculating air filter system, ideal for the direct installation in working areas with very low noise, only approx. 39 dB
- With multi-stage wideband filter
- Monitoring electronics for extraction include cool-device cable and power plug
- Optical and acoustic alarm including potential-free alarm contact
- Display with innovative touch panel
- Manual check of the filter saturation
- Eco-friendly with longer filter lifetimes and a reusable filter system
Specifications
EN 14727 | |
565 x 485 x 914 mm | |
Wing | |
6 x Shelves with sump: 25 kg/11 L each, Maximum load: 300 kg | |
Powder-coated | |
Light Gray | |
Safety storage cabinet |
6 x 25 kg/11 L PP pull-out shelves with PP sumps | |
615 x 600 x 2300 mm | |
Storage of aggressive, non-flammable hazardous materials | |
2 | |
215.4 kg | |
Steel |