missing translation for 'onlineSavingsMsg'
Learn More

SM-21 maleate, Tocris Bioscience™

Product Code. 30001461
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Presynaptic cholinergic modulator

Potent and selective σ2 antagonist with central effects following systemic administration. Causes increased release of acetylcholine at central muscarinic synapses. Potent analgesic (efficacy comparable to morphine) and nootropic agent.

Chemical Identifiers

CAS 155059-42-0
Molecular Formula C22H28ClNO7
Molecular Weight (g/mol) 453.916
InChI Key BHXGTFUQDGMXHA-NGVSCOFGSA-N
Synonym endo-8-methyl-8-azabicyclo 3.2.1 octan-3-yl 2-4-chlorophenoxy butanoate fumarate
PubChem CID 124080998
IUPAC Name (E)-but-2-enedioic acid;[(1S,5R)-8-methyl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-2-(4-chlorophenoxy)butanoate
SMILES CCC(C(=O)OC1CC2CCC(C1)N2C)OC3=CC=C(C=C3)Cl.C(=CC(=O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 453.92
Percent Purity >99%
Chemical Name or Material SM-21 maleate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'