Learn More
Scientific Industries SI™ Small Ampule/Tube Attachment
Provides vigorous end-to-end tube and ampule agitation. Scientific Industries SI™ Small Ampule/Tube Attachment accommodates up to 15 to 17mm diameter tubes or ampules.
Brand: Scientific Industries SI™ SI-0570
This item is not returnable.
View return policy
Description
- Used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse
- Tubes and ampules snap into place for vigorous end-to-end agitation
- Holds up to four 15 to 17mm diameter tubes or ampules
Specifications
Vortex-Genie 2 family of mixers, Vortex-Genie Pulse | |
4 |