Learn More
Hamilton™ Small Hub Removable Needles
Available in custom lengths and point styles to fit your laboratory needs. Hamilton™ Small Hub Removable Needles are manufactured from stainless steel.
Brand:  Hamilton™ 7731-08
 View more versions of this product
Product Code. 12316299
  • 404.00 € / Pack of 6
Explore more special offers
Description
Description
  • Autoclavable
  • Standard needle length is 2"/51mm
  • Can be used with (RN)-style syringes that are 100μL and smaller
Specifications
Specifications
Stainless Steel
0.12 mm
Removable Custom Needle
Small Removable Syringes (100μL and smaller)
23s G
0.64 mm
5 or AS